missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBA1B Monoclonal Antibody (OTI3H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506581S
This item is not returnable.
View return policy
Description
TUBA1B Monoclonal antibody specifically detects TUBA1B in Human samples. It is validated for Western BlotSpecifications
| TUBA1B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08575 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5788 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI3H2 | |
| Unconjugated | |
| PTPRC | |
| alpha tubulin; alpha-tubulin 2; Alpha-tubulin isotype M-alpha-2; alpha-tubulin ubiquitous; Detyrosinated tubulin alpha-1B chain; K-ALPHA-1; RGD1565476; Tuba1b; Tuba2; tubulin alpha 1b; tubulin alpha-1B chain; Tubulin alpha-2 chain; tubulin alpha-ubiquitous chain; Tubulin K-alpha-1; tubulin, alpha 1B; tubulin, alpha 2; tubulin, alpha, ubiquitous | |
| Full length human recombinant protein of human PTPRC produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction