missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBA6 Antibody (OTI1D8), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74692AF750
This item is not returnable.
View return policy
Description
TUBA6 Monoclonal specifically detects TUBA6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| TUBA6 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TUBA1C | |
| Full length recombinant protein of human TUBA6 (NP_116093) produced in HEK293T cells. | |
| 0.1 mL | |
| 84790.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1D8 | |
| Western Blot | |
| Alpha-tubulin 6, bcm948, MGC10851, MGC14580, TUBA6, tubulin alpha-1C chain, Tubulin alpha-6 chain, tubulin, alpha 1c, tubulin, alpha 6 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction