missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBAL3 Antibody (OTI4C5), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74696PECY7
This item is not returnable.
View return policy
Description
TUBAL3 Monoclonal specifically detects TUBAL3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.
Specifications
| TUBAL3 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| TUBAL3 | |
| Recombinant protein fragment corresponding to amino acids 150-446 of human TUBAL3 (NP_079079) produced in E.coli. | |
| 0.1 mL | |
| 79861.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI4C5 | |
| Flow Cytometry | |
| FLJ21665, MGC119347, MGC119349, tubulin alpha chain-like 3, tubulin, alpha-like 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction