missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBAL3 Monoclonal Antibody (OTI3B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503932S
This item is not returnable.
View return policy
Description
TUBAL3 Monoclonal antibody specifically detects TUBAL3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| TUBAL3 | |
| Monoclonal | |
| 0.68 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A6NHL2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79861 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI3B7 | |
| Unconjugated | |
| Tubal3 | |
| TUBAL3; Tubulin alpha chain-like 3; tubulin alpha like 3; tubulin, alpha-like 3 | |
| Human recombinant protein fragment corresponding to amino acids 150-446 of human TUBAL3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction