missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUBAL3 Monoclonal Antibody (OTI4C5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525772
This item is not returnable.
View return policy
Description
TUBAL3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.
Specifications
| TUBAL3 | |
| Monoclonal | |
| 0.55 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A6NHL2, Q3UX10 | |
| Tubal3 | |
| Human recombinant protein fragment corresponding to amino acids 150-446 of TUBAL3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI4C5 | |
| Unconjugated | |
| Tubal3 | |
| TUBAL3; Tubulin alpha chain-like 3; tubulin alpha like 3; tubulin, alpha-like 3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 238463, 291287, 487148, 79861 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction