Learn More
TUBAL3 Monoclonal Antibody (OTI5F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503911S
This item is not returnable.
View return policy
Description
TUBAL3 Monoclonal antibody specifically detects TUBAL3 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| TUBAL3 | |
| Monoclonal | |
| 1.1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14722 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7881 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| OTI5F10 | |
| Unconjugated | |
| KCNAB1 | |
| TUBAL3; Tubulin alpha chain-like 3; tubulin alpha like 3; tubulin, alpha-like 3 | |
| Full length human recombinant protein of human KCNAB1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |