missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBAL3 Monoclonal Antibody (OTI5F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503911
This item is not returnable.
View return policy
Description
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.Specifications
| TUBAL3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A6NHL2 | |
| TUBAL3 | |
| Human recombit protein fragment corresponding to amino acids 150-446 of human TUBAL3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5F10 | |
| Unconjugated | |
| TUBAL3 | |
| tubulin alpha like 3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79861 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction