missing translation for 'onlineSavingsMsg'
Learn More

Tubastatin A Hydrochloride, MedChemExpress

Product Code. 16451777
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16451777

Brand: Medchem Express HY13271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tubastatin A Hydrochloride (Tubastatin A HCl) is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay, and is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more). Tubastatin A Hydrochloride also inhibits HDAC10 and metallo-β-lactamase domain-containing protein 2 (MBLAC2).

Tubastatin A Hydrochloride (Tubastatin A HCl) is a potent and selective HDAC6 inhibitor with IC50 of 15 nM in a cell-free assay, and is selective (1000-fold more) against all other isozymes except HDAC8 (57-fold more). Tubastatin A Hydrochloride also inhibits HDAC10 and metallo-β-lactamase domain-containing protein 2 (MBLAC2).

Specifications

Chemical Name or Material Tubastatin A Hydrochloride
CAS 1310693-92-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22ClN3O2
Quantity 50 mg
Synonym Tubastatin A HCl TSA HCl
Solubility Information DMSO : 10.8 mg/mL (29.04 mM; Need ultrasonic and warming) ∣H2O : 6.67 mg/mL (17.94 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C=C1)CN2C3=C(C4=C2C=CC=C4)CN(C)CC3)NO.[H]Cl
Molecular Weight (g/mol) 371.86
Formula Weight 371.86
Percent Purity 97.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.