missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB2A Monoclonal Antibody (OTI3E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506590S
This item is not returnable.
View return policy
Description
TUBB2A Monoclonal antibody specifically detects TUBB2A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| TUBB2A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13885 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7280 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3E6 | |
| Unconjugated | |
| TUBB2A | |
| CDCBM5; class IIa beta-tubulin; dJ40E16.7; M(beta)2; TUBB; Tubb2; TUBB2A; tubb2a {ECO:0000250; tubulin beta 2A class IIa; Tubulin beta class IIa; Tubulin beta-2A chain; tubulin, beta 2A class IIA; tubulin, beta polypeptide; tubulin, beta polypeptide 2; UniProtKB:Q7TMM9} | |
| Full length human recombinant protein of human TUBB2A produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction