Learn More
Invitrogen™ TUBB2B Monoclonal Antibody (OTI6D11)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA526266
Product Code. 15935864
  • € 588.00 / 100µL
Description
TUBB2B Monoclonal Antibody for Western Blot, ICC/IF
MA1-82191 recognizes class III beta-tubulin whixh is reported to be restricted to neuronal tissue. Clone TU-20 has been used to investigate tumors of neuronal origin including neuroblastoma and ganglioneuroma. Class III beta tubulin has not been detected in tumors of non neuronal origin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TUBB2B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BVA1
TUBB2B
Full length human recombinant protein of TUBB2B produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Western Blot, Immunocytochemistry
OTI6D11
Unconjugated
TUBB2B
2410129E14Rik; bA506K6.1; beta-tubulin T beta15 (aa 1-445); brdp; class II beta-tubulin isotype; class IIb beta-tubulin; PMGYSA; RGD1309427; T beta-15; Tubb2; Tubb2a; TUBB2B; tubulin beta 2B class IIb; tubulin beta-2B chain; tubulin, beta 2; tubulin, beta 2B class IIb; tubulin, beta polypeptide paralog
Mouse
Affinity Chromatography
RUO
347733
-20°C, Avoid Freeze/Thaw Cycles