missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB2B Monoclonal Antibody (OTI8F12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506684S
This item is not returnable.
View return policy
Description
TUBB2B Monoclonal antibody specifically detects TUBB2B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| TUBB2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P54278 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5395 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI8F12 | |
| Unconjugated | |
| PMS2 | |
| 2410129E14Rik; bA506K6.1; beta-tubulin T beta15 (aa 1-445); brdp; class II beta-tubulin isotype; class IIb beta-tubulin; PMGYSA; RGD1309427; T beta-15; Tubb2; Tubb2a; TUBB2B; tubulin beta 2B class IIb; tubulin beta-2B chain; tubulin, beta 2; tubulin, beta 2B class IIb; tubulin, beta polypeptide paralog | |
| Full length human recombinant protein of human PMS2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction