missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKRF Monoclonal Antibody (OTI4C2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806097S
This item is not returnable.
View return policy
Description
NKRF Monoclonal antibody specifically detects NKRF in Human samples. It is validated for Western BlotSpecifications
| NKRF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10646 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7035 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI4C2 | |
| Unconjugated | |
| TFPI | |
| 9430034D17Rik; AW124770; C80287; ITBA4; ITBA4 protein; LOC100910054; NF-kappaB repressing factor; NF-kappa-B-repressing factor; NF-kappa-B-repressing factor-like; NFKB repressing factor; NFkB-repressing factor; NKRF; NRF; Protein ITBA4; Transcription factor NRF | |
| Human recombinant protein fragment corresponding to amino acids 29-304 of human TFPI produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction