missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TUBB6 Polyclonal Antibody

Product Code. 17743604
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17743604 20 μL 20µL
17780884 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17743604 Supplier Invitrogen™ Supplier No. PA5122206

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TUBB6 Polyclonal Antibody for Western Blot, IHC (P)

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBB6
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TUBB6
Gene Accession No. Q922F4, Q9BUF5
Gene Alias 2310057H16Rik; BB220206; class V beta-tubulin; HsT1601; TUBB-5; Tubb6; tubulin beta; tubulin beta 6 class V; tubulin beta class V; tubulin beta MGC4083; tubulin beta-6 chain; tubulin, beta 6 class V
Gene Symbols TUBB6
Host Species Rabbit
Immunogen Recombinant protein Tubulin Beta 6. The antigen corresponds to amino acid range 194-421 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307351, 67951, 84617
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.