missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUBB6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598948
This item is not returnable.
View return policy
Description
TUBB6 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.
Specifications
| TUBB6 | |
| Polyclonal | |
| Unconjugated | |
| TUBB6 | |
| 2310057H16Rik; BB220206; class V beta-tubulin; HsT1601; TUBB-5; Tubb6; tubulin beta; tubulin beta 6 class V; tubulin beta class V; tubulin beta MGC4083; tubulin beta-6 chain; tubulin, beta 6 class V | |
| Rabbit | |
| Protein G | |
| RUO | |
| 84617 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9BUF5 | |
| TUBB6 | |
| Recombinant Human Tubulin beta-6 chain protein (1-446AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction