missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubby Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97149JF646
This item is not returnable.
View return policy
Description
Tubby Polyclonal specifically detects Tubby in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Tubby | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| rd5, tubby (mouse) homolog, tubby homolog (mouse), tubby homologue, tubby protein homolog | |
| Produced in rabbits immunized with E. coli-derived Human Tubby fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuroscience, Transcription Factors and Regulators | |
| 7275 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction