Learn More
Buerkle™ Tube Cleaning Brush
Product Code. p-8003033 Shop All Bürkle Products
Click to view available options
Length (Metric):
40 cm
60 cm
80 cm
100 cm
300 cm
Diameter (Metric) Bristle:
25 mm
50 mm
Unit Size:
Each
Product Code. 10108161
Brand: Buerkle™ 53040040
Only null left
Add to basket
Add to basket
The Tube Cleaning Brush is ideal for cleaning zone samplers.
  • Manufactured from PVC and stainless steel
  • Available in a range of lengths
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Brush
Diameter (Metric) Bristle 25 mm
For Use With (Application) Used for Cleaning
For Use With (Equipment) Zone Sampler
Length (Metric) 40 cm
Material PVC, Stainless Steel