Learn More
Carbolite™ Tube Furnace
Click to view available options
Depth (Metric):
175 mm
375 mm
Unit Size:
Each
Specifications
Specifications
| Analysis Time | 15 min. |
| Type | Tube Furnace |
| Includes | Standard Controller |
| Temperature Stability | 1200°C |
| Dimensions (L x W x H) Exterior | 370 W x 375 H mm |
| Length (Metric) | 300 mm |
| Width (Metric) | 370 mm |
| Height (Metric) | 375 mm |
| Depth (Metric) | 375 mm |