Learn More
OHAUS™ Tube Holder
OHAUS™ Tube Holder is constructed of polyethylene foam and can be used in a range of applications.
Brand: OHAUS™ 30400231
Additional Details : Weight : 0.90000kg
Description
- Can be used on all vortex mixers
- Increase throughput by mixing multiple samples at one time
- Constructed of polyethylene foam
- Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
Specifications
20 to 25 mm Tubes | |
Tube Holder | |
Holder For 20-25 mm Tubes, Pack of 2 | |
2/Pk. |
20 to 22 mm | |
Foam | |
8 |