Learn More
OHAUS™ Tube Holder
Product Code. 15762619 Shop All Ohaus Products
Click to view available options
Holds:
14 to 19 mm Tubes
18 x 0.5 mL Microtubes
20 to 25 mm Tubes
24 x 1.5 to 2 mL Microtubes
9 to 13 mm Tubes
Microtubes
Unit Size:
Each
Pack of 2
Product Code. 15762619
Brand: OHAUS™ 30400204
Only null left
Add to basket
Add to basket
OHAUS™ Tube Holder is constructed of polyethylene foam and can be used in a range of applications.
  • Can be used on all vortex mixers
  • Increase throughput by mixing multiple samples at one time
  • Constructed of polyethylene foam
  • Choose the appropriate holder for the sample size, or select the flat insert and cut out to suit custom specifications
TRUSTED_SUSTAINABILITY
Specifications
Holds 18 x 0.5 mL Microtubes
Product Type Tube Holder
Material Foam
Description 0.5 mL Micro-Tube Holder
No. of Positions 18