missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph instruments Tube MonoBlock
Accommodate specifically sized reagent tubes at your StarFish magnetic stirrer workstation. Heidolph™ Tube MonoBlock, constructed of one piece, is ideal for experiments using the same vessel.
€ 535.00 - € 540.00
Specifications
Description | MonoBlock |
---|---|
For Use With (Equipment) | Starfish Workstation, MR-Hei Magnetic Stirrers |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11751699
|
Heidolph instruments
505-80004-00 |
40 x 16 mm Tubes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11731699
|
Heidolph instruments
505-80002-00 |
16 x 25 mm Tubes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11741699
|
Heidolph instruments
505-80003-00 |
16 x 24 mm Tubes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11761699
|
Heidolph instruments
505-80005-00 |
40 x 12 mm Tubes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Accommodates one specific tube size.
- One-piece construction.
Specifications
MonoBlock | |
Starfish Workstation, MR-Hei Magnetic Stirrers |