Learn More
Cytiva Tube Rack
Shop All Cytiva Products
Click to view available options
Holds:
Tubes (50 mL)
Unit Size:
Each
Description
Holds up to 55 tubes. For use with Fraction collector F9-C and ÅKTA avant.
Specifications
Specifications
| Product Type | Tube Rack |
| Holds | Tubes (50 mL) |
| Positions | 55 |
| Quantity | 1 Each |