missing translation for 'onlineSavingsMsg'
Learn More

Benchmark Scientific™ Tube Rack for BenchMixer XL and XLQ

Product Code. 17984402
Click to view available options
Holds:
15 x 25 mm Tubes
50 x 13 mm Tubes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17984402

Brand: Benchmark Scientific™ BV101013

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This foam rack holds test tubes and is designed for use with the BenchMixer XL and XLQ.

  • Foam rack accommodates test tubes
  • Fits BenchMixer XL and XLQ

Specifications

Holds 50 x 13 mm Tubes
For Use With (Application) Vortexing of 13 mm Diameter Tubes
For Use With (Equipment) BenchMixer XL and XLQ
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.