missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ Tube Strip Rack Set

Product Code. 12715204
Change view
Click to view available options
:
Quantity: 4 Sets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12715204 Quantity: 4 Sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12715204 Supplier Macherey-Nagel Bioanalysis™ Supplier No. 740477
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Tube Strip Rack Set that includes 1 rack, 12 strips with 8 tubes, and 12 cap strips.

  • Set consists of a rack, 12 strips with 8 tubes each, and 12 cap strips
  • Includes 4 sets

Specifications

Format Strip Plate
Content And Storage 18°C to 25°C
No. of Wells 8
Quantity 4 Sets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.