missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TUBE1 Polyclonal Antibody

Product Code. 15754821 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15754821 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15754821 Supplier Invitrogen™ Supplier No. PA556917

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: WNQEGWKTSL CSVPPVGHSH SLLALANNTC VKPTFMELKE RFMRLYKKKA HLHHYLQVEG MEESCFTEAV SSLSALIQEY DQLDATKNMP VQDLPRLS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

Microtubules constitute one of the major components of eukaryotic cells cytoskeleton and are involved in many essential processes, including cell division, ciliary and flagellar motility and intracellular transport. Microtubules of the eukaryotic cytoskeleton are composed of a heterodimer of α- and β-tubulin. In addition to α- and β-tubulin, several other tubulins have been identified, bringing the number of distinct tubulin classes to seven. Most of these tubulins have distinct subcellular localization and an emerging, diverse set of functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUBE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TUBE1
Gene Accession No. Q9UJT0
Gene Alias 2310061K05Rik; AI551343; dJ142L7.2; epsilon-tubulin; epsilon-tubulin 1; FLJ22589; FLJ44203; RP1-142L7.1; TUBE; Tube1; Tubulin; tubulin epsilon 1; tubulin epsilon chain; tubulin, epsilon 1
Gene Symbols TUBE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TUBE1. Recombinant protein control fragment (Product #RP-94619).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51175
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.