missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TUBE1 Polyclonal Antibody
Description
Antibody detects endogenous levels of total TUBE1.
Microtubules constitute one of the major components of eukaryotic cells cytoskeleton and are involved in many essential processes, including cell division, ciliary and flagellar motility and intracellular transport. Microtubules of the eukaryotic cytoskeleton are composed of a heterodimer of α- and β-tubulin. In addition to α- and β-tubulin, several other tubulins have been identified, bringing the number of distinct tubulin classes to seven. Most of these tubulins have distinct subcellular localization and an emerging, diverse set of functions.
Specifications
Specifications
| Antigen | TUBE1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TUBE1 |
| Gene Accession No. | Q9D6T1, Q9UJT0 |
| Gene Alias | 2310061K05Rik; AI551343; dJ142L7.2; epsilon-tubulin; epsilon-tubulin 1; FLJ22589; FLJ44203; RP1-142L7.1; TUBE; Tube1; Tubulin; tubulin epsilon 1; tubulin epsilon chain; tubulin, epsilon 1 |
| Gene Symbols | TUBE1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?