Learn More
Terumo™ Tuberculin Syringe
Click to view available options
Needle Gauge:
25 G
Unit Size:
Pack of 100
Description
- Ultra-sharp needles
- Clear barrels
- Bold, precise scale markings
- Smooth plunger action
- Tamper evident peel packs
Specifications
Specifications
| Certifications/Compliance | BD |
| Needle Gauge | 25 G |
| For Use With (Application) | Tuberculin |
| Length (Metric) | 160 mm |
| Product Type | Tuberculin Syringe |
| Quantity | 100/Pk. |
| Volume (Metric) | 1 mL |
| Connection Type | Needle |