missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBGCP5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94420-0.02ml
This item is not returnable.
View return policy
Description
TUBGCP5 Polyclonal antibody specifically detects TUBGCP5 in Rat samples. It is validated for Western Blot
Specifications
| TUBGCP5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| gamma-tubulin complex component 5, gamma-tubulin complex component GCP5, GCP5KIAA1899GCP-5, tubulin, gamma complex associated protein 5 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TUBGCP5 (NP_443135.3). KEIKTDAHYSILSLLLCLSDSPSNSSYVETPRNKEVEKKDDFDWGKYLMEDEEMDIGPYMDTPNWSEESEEENDQQPLSREDSGIQVDRTPLEEQDQNRKL | |
| 0.02 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 114791 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction