Learn More
Masterflex™ Tubing Adapter NPT (F) Polypropylene
Ideal for general-purpose and single-use applications, to connect tubing with rest of system
Brand:  Masterflex™ 31200-10
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11745978
  • € 161.00 / Each
Estimated Shipment: 20-09-2024
to see stock.
Add to basket
Description
Description
  • Determine which connector type is needed for your application by contacting free technical application assistance
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp™ connections
  • Excellent chemical resistance; low leachable characteristics
Specifications
Specifications
Vacuum Tubing
Masterflex L/S and I/P sanitary tubing connection
1/8 in.
Tubing Adapter; Polypropylene (PP), 3/4 in. mini to 1/8 in. NPT (F)
Sanitary x NPT(F)