Learn More
Biochrom Tubing for Automatic Sipper
Click to view available options
Type:
Standard Tubing
Unit Size:
Each
Description
- Includes:
- 6 marprene pump head tubes
- PTFE tubing
- Silicon connection tubing
- For use with Ultrospec 7000/8000/9000 sipper
- Easy to install
Specifications
Specifications
| Type | Standard Tubing |
| For Use With (Application) | Spectrophotometry |
| For Use With (Equipment) | Ultrospec 7000/8000/9000 Spectrophotometer Sipper |
| Includes | 6 Marprene Pump Head Tubes, PTFE Tubing, Silicon Connection Tubing |