missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin alpha-1B Antibody (OTI3G3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74703AF350
This item is not returnable.
View return policy
Description
Tubulin alpha-1B Monoclonal specifically detects Tubulin alpha-1B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Tubulin alpha-1B | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TUBA1B | |
| Full length recombinant protein of human Tubulin alpha-1B (NP_006073) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI3G3 | |
| Western Blot, Immunohistochemistry | |
| alpha tubulin, Alpha-tubulin ubiquitous, K-ALPHA-1, tubulin alpha-1B chain, Tubulin alpha-ubiquitous chain, Tubulin K-alpha-1, tubulin, alpha 1b, tubulin, alpha, ubiquitous | |
| Mouse | |
| 0.1 mL | |
| 10376 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction