missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin alpha-3E Antibody (OTI2A8), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74704IR
This item is not returnable.
View return policy
Description
Tubulin alpha-3E Monoclonal specifically detects Tubulin alpha-3E in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| Tubulin alpha-3E | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TUBA3E | |
| Full length recombinant protein of human Tubulin alpha-3E (NP_997195) produced in HEK293T cells. | |
| 0.1 mL | |
| 112714.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI2A8 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Alpha-tubulin 3E, tubulin alpha-3E chain, tubulin, alpha 3e | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction