Learn More
WFDC5 Antibody (M47-P2D5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50363AF647
Product Code. 18156009
  • € 437.00 / 0.10mL
Description
WFDC5 Monoclonal specifically detects WFDC5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WFDC5
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
WFDC5
Ovalbumin-conjugated synthetic peptide TERVHDGRPG
0.1 mL
Cancer, Cellular Signaling
149708
Store at 4C in the dark.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry
M47-P2D5
Western Blot, ELISA, Immunohistochemistry
dJ211D12.5, PRG5protease inhibitor WAP1, Putative protease inhibitor WAP1, WAP four-disulfide core domain 5, WAP four-disulfide core domain protein 5, WAP1p53-responsive gene 5 protein