missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin alpha-8 Antibody (OTI2G6), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74709AF594
This item is not returnable.
View return policy
Description
Tubulin alpha-8 Monoclonal specifically detects Tubulin alpha-8 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Tubulin alpha-8 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TUBA8 | |
| Full length recombinant protein of human Tubulin alpha-8 (NP_061816) produced in HEK293T cells. | |
| 0.1 mL | |
| 51807.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2G6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Alpha-tubulin 8, TUBAL2Tubulin alpha chain-like 2, tubulin alpha-8 chain, tubulin, alpha 8 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction