missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tubulin beta-2C Polyclonal Antibody
GREENER_CHOICE

Product Code. 17287663 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17287663 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17287663 Supplier Invitrogen™ Supplier No. PA5114903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total beta II Tubulin.

Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tubulin beta-2C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TUBB4B
Gene Accession No. P68371, P68372, Q6P9T8
Gene Alias 4930542G03Rik; Beta2; class IVb beta tubulin; class IVb beta-tubulin; LOW QUALITY PROTEIN: tubulin beta chain; novel tubulin protein; Tubb2; TUBB2C; Tubb2c1; TUBB4B; tubulin beta; tubulin beta 4B class IVb; tubulin beta-2 chain; Tubulin beta-2C chain; tubulin beta-4B chain; tubulin, beta 2C; tubulin, beta 2c1; tubulin, beta 4B class IVb; tubulin, beta, 2
Gene Symbols TUBB4B
Host Species Rabbit
Immunogen A synthesized peptide derived from human TUBB4B(Accession P68371), corresponding to amino acid residues E410-A445.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10383, 227613, 296554
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.