missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tubulin Beta 4 Antibody (OTI3F1), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74713DL594
This item is not returnable.
View return policy
Description
Tubulin Beta 4 Monoclonal antibody specifically detects Tubulin Beta 4 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Proximity Ligation Assay
Specifications
| Tubulin Beta 4 | |
| Monoclonal | |
| DyLight 594 | |
| TUBB5beta-5, Tubulin 5 beta, tubulin beta-4 chain, tubulin, beta 4, tubulin, beta, 5 | |
| Full length recombinant protein of human Tubulin Beta 4 (NP_006078) produced in HEK293T cells. | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cellular Markers | |
| 10382 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Proximity Ligation Assay | |
| OTI3F1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction