missing translation for 'onlineSavingsMsg'
Learn More

TUG-891, MedChemExpress

Product Code. 16453208
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453208 10 mg 10mg
16465937 5 mg 5mg
16449756 50 mg 50mg
16432676 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16453208 Supplier Medchem Express Supplier No. HY10088110mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TUG-891 is a potent and selective agonist for the long chain free fatty acid (LCFA) receptor 4 (FFA4/GPR120).

TUG-891 is a potent and selective agonist for the long chain free fatty acid (LCFA) receptor 4 (FFA4/GPR120).

Specifications

Chemical Name or Material TUG-891
CAS 1374516-07-0
Color White
Purity Grade Notes Research
Molecular Formula C23H21FO3
Quantity 10 mg
Solubility Information DMSO : ≥ 180 mg/mL (493.95 mM)
SMILES O=C(O)CCC1=CC=C(OCC2=CC(F)=CC=C2C3=CC=C(C)C=C3)C=C1
Molecular Weight (g/mol) 364.41
Formula Weight 364.41
Percent Purity 99.36%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.