missing translation for 'onlineSavingsMsg'
Learn More

TUG Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-28710G

Product Code. 18490006

  • € 633.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TUG Polyclonal antibody specifically detects TUG in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TUG
Polyclonal
Western Blot
Alveolar soft part sarcoma chromosomal region candidate gene 1 protein, alveolar soft part sarcoma chromosome region, candidate 1, Alveolar soft part sarcoma locus, ASPLUBX domain-containing protein 9, ASPSrenal cell carcinoma gene on chromosome 17, RCC17renal cell carcinoma, papillary, 17, Renal papillary cell carcinoma protein 17, tether containing UBX domain for GLUT4, TUG, UBX domain protein 9, UBXD9ASPCR1, UBXN9FLJ45380
The immunogen recognized by this antibody maps to a region between residue 503 and 553 of human alveolar soft part sarcoma locus using the numbering given in entry NP_076988.1 (GeneID 79058).
0.1 mL
Cancer, Cell Biology, Endocrinology, Signal Transduction
79058
Store at 4°C in the dark.
IgG
Western Blot
DyLight 488
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.