missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TULP3 Antibody (OTI7B8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74721B
This item is not returnable.
View return policy
Description
TULP3 Monoclonal specifically detects TULP3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TULP3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TULP3 | |
| Full length recombinant protein of human TULP3 (NP_003315) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI7B8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| MGC45295, tubby like protein 3, tubby-related protein 3, TUBL3Tubby-like protein 3 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 7289.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction