Learn More
TULP3 Monoclonal Antibody (OTI6H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504150
This item is not returnable.
View return policy
Description
This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.Specifications
| TULP3 | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75386 | |
| TULP3 | |
| Full length human recombit protein of human TULP3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI6H4 | |
| Unconjugated | |
| TULP3 | |
| TUBL3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7289 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |