Learn More
TULP3 Mouse anti-Canine, Human, Mouse, Clone: OTI1A10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504148
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.Specifications
| TULP3 | |
| Monoclonal | |
| Unconjugated | |
| TULP3 | |
| TUBL3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 22158, 611832, 7289 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1A10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O75386, O88413 | |
| TULP3 | |
| Full length human recombit protein of human TULP3 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.