Learn More
RETSCH Tungsten Carbide Grinding Sets
Product Code. 15320974 Shop All Retsch Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15320974 50 mL Each
15340974 250 mL Each
15330974 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15320974 Supplier RETSCH Supplier No. 01.462.0353
Only null left
Specifications
Product Type Grinding Set
Capacity (Metric) 50 mL
For Use With (Equipment) For RS200/RS100
Material Tungsten Carbide
Speed 1200 rpm