Learn More
Heathrow Scientific™ Tungsten Point Lab Pen
Etches glass, plastic, and ceramic. Heathrow Scientific™ Tungsten Point Lab Pen contains a sharp, tungsten carbide tip to permaently mark slides, test tubes, and labware.
Brand:  Heathrow Scientific™ HS1846
Additional Details : Weight : 0.00910kg
Product Code. 12397559
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Retractable tip for safe storage
  • Measures 9.8mm dia. x 137mmL
Specifications
Specifications
Pen
Tungsten Carbide
Etching glass, ceramic, and plastic
Retractable