Learn More
Honeywell™ Turboshield Hard Hat Adapter
Click to view available options
:
For use with hard hats, Turboshield face shield
Description
- Features a patented dual-pivot mechanism and a patented visor latch mechanism release system
- Perfect for workers exposed to falling for flying objects, impact, splash, UV and airborne debris
- Hard hat adapter option makes it easier for those who need to combine head protection with face protection
- Visors slide back 170 mm, improving balance and weight distribution
- Dual-hinge design increases clearance by as much as 100 mm and keeps accessory slots open for compatibility with other PPE
- Compatible with Supervisor specialty visors including gold-coated, triacetate and acetate
- Compatible with most major hard hats;(including North brand) with the exception of full brim hard hats.
Specifications
Specifications
| Certifications/Compliance | EC Category PPE: 2, EC Certificate: C1519.2HON, EC Attestation: C1519.2HON, EN 166 |
| Description | Dielectric |
| For Use With (Equipment) | Hard hats, Turboshield face shield |
| For Use With (Application) | Impact protection, splash protection, UV protection |
| Frame Color | Gray |
| Frame Material | Reinforced Nylon |