missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TUSC2 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
:
0.1 mL
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1M Urea, pH 7.4. |
| Gene ID (Entrez) | 11334 |
| Molecular Weight (g/mol) | 29 kDa |
| Quantity | 0.1 mL |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | C3orf11PDAP2, Fus-1 protein, FUS1Fusion 1 protein, LGCC, PAP, PDGFA associated protein 2, PDGFA-associated protein 2, tumor suppressor candidate 2 |
| Gene Symbol | TUSC2 |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-86794. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL. |
| Research Category | Cell Cycle and Replication |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction