Learn More
TUSC3 Mouse anti-Human, Unconjugated, Clone: 1D3G12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67382-1-IG-20UL
 View more versions of this product
Product Code. 16899975
  • € 145.00 / 20µL
Description
The TUSC3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TUSC3. This antibody recognizes human antigen. The TUSC3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TUSC3
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1D3G12
Unconjugated
20 μL
7991