missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWA1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77366R
This item is not returnable.
View return policy
Description
TWA1 Polyclonal antibody specifically detects TWA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| TWA1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 54994 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| bA305P22.1, C20orf11, chromosome 20 open reading frame 11, FLJ20602, GID complex subunit 8 homolog (S. cerevisiae), Twa1, TWA1protein C20orf11, two hybrid associated protein 1, Two hybrid-associated protein 1 with RanBPM | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human TWA1. The immunogen is located within the last 50 amino acids of TWA1. Amino Acid Squence: NELDQKKVKYPKMTDL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction