missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK R/TNFRSF12 Antibody (314636), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB16101MFV610
This item is not returnable.
View return policy
Description
TWEAK R/TNFRSF12 Monoclonal antibody specifically detects TWEAK R/TNFRSF12 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| TWEAK R/TNFRSF12 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 314636 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
| E. coli-derived recombinant mouse TWEAK R, Glu28-Trp79, Accession # Q9CR75.1 | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors | |
| 51330 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction