missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK R/TNFRSF12 Antibody (ITEM-4), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00409PECY55
This item is not returnable.
View return policy
Description
TWEAK R/TNFRSF12 Monoclonal specifically detects TWEAK R/TNFRSF12 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| TWEAK R/TNFRSF12 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| TNFRSF12A | |
| The immunogen for this antibody is TWEAKR. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| ITEM-4 | |
| Flow Cytometry | |
| CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14 | |
| Mouse | |
| Protein A or G purified | |
| Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors | |
| 51330.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction