Learn More
TWEAK/TNFSF12 Antibody (C06-3C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11625-0.025mg
 View more versions of this product
Product Code. 18631719
  • € 203.00 / 25µg
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TWEAK/TNFSF12
Monoclonal
Unconjugated
APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12
E. coli derived recombinant protein corresponding to aa 97-249 of human TWEAK
0.025 mg
Angiogenesis, Apoptosis, Cancer, Tumor Suppressors
8742
Store at -20°C. Avoid freeze-thaw cycles.
IgG1
ELISA
C06-3C8
PBS
Mouse
Protein G purified