missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TWEAK/TNFSF12 Antibody (MTW-1) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
€ 250.00 - € 680.00
Specifications
| Antigen | TWEAK/TNFSF12 |
|---|---|
| Clone | MTW-1 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622399
|
Novus Biologicals
NBP3-09046-0.2mg |
0.2 mg |
€ 680.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668837
|
Novus Biologicals
NBP3-09046-0.025mg |
0.025 mg |
€ 250.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Mouse samples. It is validated for Flow Cytometry, NeutralizationSpecifications
| TWEAK/TNFSF12 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| This antibody was raised by immunising SD rats with CD8-mTWEAK, a fusion of the human CD8alpha extracellular region to murine TWEAK. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| MTW-1 | |
| Flow Cytometry, Neutralization | |
| Unconjugated | |
| Rat | |
| Angiogenesis, Apoptosis, Cancer, Tumor Suppressors | |
| PBS | |
| 8742 | |
| IgG1 κ | |
| Protein A purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto